Product Details
- SNP ID
-
rs74965018
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:26046840 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCCCTGAGTTGGCGCTGGAGCT[G/T]GAACTGGCAGCTGTGCTGGACTGGC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SHISA2
PubMed Links
Gene Details
- Gene
- SHISA2
- Gene Name
- shisa family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007538.1 |
646 |
Silent Mutation |
TCA,TCC |
S187S |
NP_001007539.1 |
View Full Product Details