Product Details

SNP ID
rs75698534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27436031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGATTAGCTAATCAGTTCATAC[A/G]TATCTGGTTAAGTTGACTGAAATCT
Phenotype
MIM: 600860
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF3A PubMed Links

Gene Details

Gene
GTF3A
Gene Name
general transcription factor IIIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002097.2 Intron NP_002088.2
Gene
MTIF3
Gene Name
mitochondrial translational initiation factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166261.1 Intron NP_001159733.1
NM_001166262.1 Intron NP_001159734.1
NM_001166263.1 Intron NP_001159735.1
NM_152912.4 Intron NP_690876.3
XM_006719771.3 Intron XP_006719834.1
XM_006719772.3 Intron XP_006719835.1
XM_011534957.2 Intron XP_011533259.1
XM_011534958.2 Intron XP_011533260.1
XM_011534959.2 Intron XP_011533261.1
XM_011534960.2 Intron XP_011533262.1
XM_011534961.2 Intron XP_011533263.1
XM_011534962.2 Intron XP_011533264.1
XM_011534963.2 Intron XP_011533265.1
XM_017020417.1 Intron XP_016875906.1

View Full Product Details