Product Details

SNP ID
rs75239949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:76360231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAAAGGCCAACTGCACTTTTCC[A/G]TGCCGCTTCTGAAACAGTAAAAATG
Phenotype
MIM: 615114
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF516 PubMed Links

Gene Details

Gene
ZNF516
Gene Name
zinc finger protein 516
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014643.3 6028 UTR 3 NP_055458.1
XM_011526269.1 6028 UTR 3 XP_011524571.1
XM_011526270.2 6028 UTR 3 XP_011524572.1
XM_011526271.2 6028 UTR 3 XP_011524573.1
XM_011526272.2 6028 UTR 3 XP_011524574.1
XM_011526273.2 6028 UTR 3 XP_011524575.1
XM_011526274.2 6028 UTR 3 XP_011524576.1
XM_011526275.2 6028 UTR 3 XP_011524577.1
XM_011526276.1 6028 UTR 3 XP_011524578.1
XM_011526277.2 6028 Intron XP_011524579.1
XM_017026097.1 6028 UTR 3 XP_016881586.1

View Full Product Details