Product Details

SNP ID
rs76177820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:57355655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTAATTTTATGGACAACCCAGAA[C/T]GTTTTCATGGGTAGAACACATTCTA
Phenotype
MIM: 609478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST8SIA3 PubMed Links

Gene Details

Gene
ST8SIA3
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015879.2 Intron NP_056963.2

View Full Product Details