Product Details

SNP ID
rs80289950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63523827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACCATCCCAGCCAAGCAAAAAG[C/T]GGTACTTTCTCAAGGACAGAAACAG
Phenotype
MIM: 615662
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINB12 PubMed Links

Gene Details

Gene
SERPINB12
Gene Name
serpin family B member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307928.1 Intron NP_001294857.1
NM_080474.2 Intron NP_536722.1
XM_005266778.3 Intron XP_005266835.2
XM_011526246.1 Intron XP_011524548.1
XM_011526248.1 Intron XP_011524550.1
XM_011526249.1 Intron XP_011524551.1
XM_017026067.1 Intron XP_016881556.1

View Full Product Details