Product Details

SNP ID
rs79551794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:64082622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTAACCAGGAATTCACATCACTG[A/G]CTTGCTGACAGTGTGAATTCTGTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00305 PubMed Links

Gene Details

Gene
LINC00305
Gene Name
long intergenic non-protein coding RNA 305
There are no transcripts associated with this gene.

View Full Product Details