Product Details

SNP ID
rs79008323
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109979631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGCTACCTGTCCCTGTATAACGC[G/T]CTCACCTATTACTCGCGCCGGACCC
Phenotype
MIM: 600553
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
GPR6 PubMed Links

Gene Details

Gene
GPR6
Gene Name
G protein-coupled receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286099.1 858 Silent Mutation GCG,GCT A188A NP_001273028.1
NM_005284.4 858 Silent Mutation GCG,GCT A173A NP_005275.1

View Full Product Details