Product Details
- SNP ID
-
rs75938999
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:110246118 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTACCAGAACCGCACAACGCTGCTG[C/T]CACTGCCACTGACCTCAAACCCAGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
METTL24
PubMed Links
Gene Details
- Gene
- METTL24
- Gene Name
- methyltransferase like 24
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001123364.1 |
929 |
Missense Mutation |
GAC,GGC |
D310G |
NP_001116836.1 |
View Full Product Details