Product Details

SNP ID
rs78567928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:96896800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATACACATCTTTTAGAAACGACA[A/G]CAGCTTTTCATCTTTACGAGCAATC
Phenotype
MIM: 611776
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL32 PubMed Links

Gene Details

Gene
KLHL32
Gene Name
kelch like family member 32
There are no transcripts associated with this gene.

Gene
NDUFAF4
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014165.3 Intron NP_054884.1

View Full Product Details