Product Details

SNP ID
rs75032644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116461964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCGGGCGGTGCTGGACCTGCACG[G/T]CAAGCACCACAGCGCCTTGGGCTAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM26F PubMed Links

Gene Details

Gene
FAM26F
Gene Name
family with sequence similarity 26 member F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010919.2 437 Missense Mutation GGC,GTC G12V NP_001010919.1
NM_001276460.1 437 Intron NP_001263389.1
XM_005266999.3 437 Missense Mutation GGC,GTC G12V XP_005267056.1
XM_011535845.2 437 Missense Mutation GGC,GTC G12V XP_011534147.1

View Full Product Details