Product Details

SNP ID
rs76802220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116511791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTGCTGACCGTGGGAAGTGAGC[A/G]TCTCTTTTCTGTTGTGGCTTTTAAG
Phenotype
MIM: 614137
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM26E PubMed Links

Gene Details

Gene
FAM26E
Gene Name
family with sequence similarity 26 member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153711.3 147 Missense Mutation CAT,CGT H32R NP_714922.1
Gene
TRAPPC3L
Gene Name
trafficking protein particle complex 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139444.2 147 Intron NP_001132916.1

View Full Product Details