Product Details

SNP ID
rs78445627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116752339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGAAGTGTTGCCAATATTCTAC[A/G]TCTTCACTTTGGCAGTGGTTACACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM162B PubMed Links

Gene Details

Gene
FAM162B
Gene Name
family with sequence similarity 162 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085480.2 894 UTR 3 NP_001078949.1

View Full Product Details