Product Details

SNP ID
rs75882246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138425724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCCTGTACTAAGATGTACCCCA[C/T]TCCAGGTCCTAGAACAACTCCTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR3145 PubMed Links

Gene Details

Gene
MIR3145
Gene Name
microRNA 3145
There are no transcripts associated with this gene.

Gene
NHSL1
Gene Name
NHS like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144060.1 Intron NP_001137532.1
NM_020464.1 Intron NP_065197.1
XM_005267062.4 Intron XP_005267119.1
XM_011535965.2 Intron XP_011534267.1
XM_011535966.2 Intron XP_011534268.1
XM_011535967.2 Intron XP_011534269.1
XM_011535968.2 Intron XP_011534270.1
XM_011535969.2 Intron XP_011534271.1
XM_011535970.2 Intron XP_011534272.1
XM_011535971.2 Intron XP_011534273.1
XM_011535972.2 Intron XP_011534274.1
XM_011535973.2 Intron XP_011534275.1
XM_011535974.2 Intron XP_011534276.1
XM_011535976.1 Intron XP_011534278.1
XM_017011086.1 Intron XP_016866575.1
XM_017011087.1 Intron XP_016866576.1
XM_017011088.1 Intron XP_016866577.1
XM_017011089.1 Intron XP_016866578.1

View Full Product Details