Product Details

SNP ID
rs78275140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143495779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGATGAATGGTTAGCTGTGGCAG[G/T]TCTACCATAATGCCATTTTGCTCCA
Phenotype
MIM: 136820 MIM: 603164
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FUCA2 PubMed Links

Gene Details

Gene
FUCA2
Gene Name
fucosidase, alpha-L- 2, plasma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032020.4 1581 Missense Mutation GAA,GAC E444D NP_114409.2
Gene
PEX3
Gene Name
peroxisomal biogenesis factor 3
There are no transcripts associated with this gene.

View Full Product Details