Product Details

SNP ID
rs75667067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:150874086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATAAGAACCCAGCATAGCCAAA[G/T]TAACAGTCACGACAGGCTGGAAAGA
Phenotype
MIM: 611427
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MTHFD1L PubMed Links

Gene Details

Gene
MTHFD1L
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242767.1 Intron NP_001229696.1
NM_001242768.1 Intron NP_001229697.1
NM_001242769.1 Intron NP_001229698.1
NM_015440.4 Intron NP_056255.2
XM_005266907.4 Intron XP_005266964.1
XM_005266910.4 Intron XP_005266967.1
XM_005266911.4 Intron XP_005266968.1
XM_011535729.2 Intron XP_011534031.1
XM_011535730.2 Intron XP_011534032.1
XM_011535731.2 Intron XP_011534033.1
XM_011535732.2 Intron XP_011534034.1
XM_011535733.2 Intron XP_011534035.1
XM_011535734.2 Intron XP_011534036.1
XM_011535737.2 Intron XP_011534039.1
XM_011535738.2 Intron XP_011534040.1
XM_017010702.1 Intron XP_016866191.1
XM_017010703.1 Intron XP_016866192.1
XM_017010704.1 Intron XP_016866193.1
XM_017010705.1 Intron XP_016866194.1
XM_017010706.1 Intron XP_016866195.1
XM_017010707.1 Intron XP_016866196.1
XM_017010708.1 Intron XP_016866197.1

View Full Product Details