Product Details
- SNP ID
-
rs77783612
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:157719672 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGATTAGAGGAGAGACACACACAC[C/T]GTAGCTCAGTGATGCTCAAAGGAAG
- Phenotype
-
MIM: 605952
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNX9
PubMed Links
Gene Details
- Gene
- SNX9
- Gene Name
- sorting nexin 9
View Full Product Details