Product Details
- SNP ID
-
rs75725528
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:169789063 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCAGGACACATGAGGCACATATG[A/T]GCTGCTGTGCATTCCTCAGTGAATA
- Phenotype
-
MIM: 615532
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ERMARD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139182323] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ERMARD
- Gene Name
- ER membrane associated RNA degradation
There are no transcripts associated with this gene.
- Gene
- LINC00242
- Gene Name
- long intergenic non-protein coding RNA 242
There are no transcripts associated with this gene.
- Gene
- LINC00574
- Gene Name
- long intergenic non-protein coding RNA 574
There are no transcripts associated with this gene.
View Full Product Details