Product Details

SNP ID
rs74456177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20117734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGACCCTTGCTTCTTTTGCCACC[C/T]CTAAGGTGATTCTGAACTTGTTAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4K17 PubMed Links

Gene Details

Gene
OR4K17
Gene Name
olfactory receptor family 4 subfamily K member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004715.1 328 Missense Mutation CCT,TCT P110S NP_001004715.1

View Full Product Details