Product Details

SNP ID
rs79751013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35541653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCACTGCAGATGGATGGCAGAAG[G/T]AAAGTGAAGAGCAGTAGTAGCTCAT
Phenotype
MIM: 614027 MIM: 608884
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
INSM2 PubMed Links

Gene Details

Gene
INSM2
Gene Name
INSM transcriptional repressor 2
There are no transcripts associated with this gene.

Gene
RALGAPA1
Gene Name
Ral GTPase activating protein catalytic alpha subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283043.1 Intron NP_001269972.1
NM_001283044.1 Intron NP_001269973.1
NM_014990.1 Intron NP_055805.1
NM_194301.2 Intron NP_919277.2
XM_005267491.3 Intron XP_005267548.1
XM_005267492.3 Intron XP_005267549.1
XM_005267493.3 Intron XP_005267550.1
XM_005267498.4 Intron XP_005267555.1
XM_006720098.2 Intron XP_006720161.1
XM_006720099.2 Intron XP_006720162.1
XM_006720100.3 Intron XP_006720163.1
XM_006720101.2 Intron XP_006720164.1
XM_006720102.3 Intron XP_006720165.1
XM_006720103.2 Intron XP_006720166.1
XM_006720104.3 Intron XP_006720167.1
XM_011536615.2 Intron XP_011534917.1
XM_011536616.2 Intron XP_011534918.1
XM_017021141.1 Intron XP_016876630.1
XM_017021142.1 Intron XP_016876631.1
XM_017021143.1 Intron XP_016876632.1
XM_017021144.1 Intron XP_016876633.1
XM_017021145.1 Intron XP_016876634.1
XM_017021146.1 Intron XP_016876635.1
XM_017021147.1 Intron XP_016876636.1

View Full Product Details