Product Details

SNP ID
rs75463308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20510729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTGGCCAGAAGATCCCATCCT[C/G]GGTGAAGATGAGGTTGGGGGTAACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNASE10 PubMed Links

Gene Details

Gene
RNASE10
Gene Name
ribonuclease A family member 10 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012975.1 258 Silent Mutation CTC,CTG L86L NP_001012993.1

View Full Product Details