Product Details

SNP ID
rs80073712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20739616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTTGTGTTGCTCTGCATCCTCA[C/T]CAGCTTTGGTGATGTCAGTTTGTAT
Phenotype
MIM: 611580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EDDM3A PubMed Links

Gene Details

Gene
EDDM3A
Gene Name
epididymal protein 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006683.4 Intron NP_006674.2
XM_017020934.1 Intron XP_016876423.1

View Full Product Details