Product Details

SNP ID
rs78315193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21033265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGAAACCATGATTTCACAGAG[A/G]GAAGCAGGCTGGAAACTGGGGGTTG
Phenotype
MIM: 605272 MIM: 612484 MIM: 616956
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6717 PubMed Links

Gene Details

Gene
MIR6717
Gene Name
microRNA 6717
There are no transcripts associated with this gene.

Gene
NDRG2
Gene Name
NDRG family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282211.1 1162 Intron NP_001269140.1
NM_001282212.1 1162 Intron NP_001269141.1
NM_001282213.1 1162 Intron NP_001269142.1
NM_001282214.1 1162 Intron NP_001269143.1
NM_001282215.1 1162 Intron NP_001269144.1
NM_001282216.1 1162 Intron NP_001269145.1
NM_001320329.1 1162 Intron NP_001307258.1
NM_016250.2 1162 Intron NP_057334.1
NM_201535.1 1162 Intron NP_963293.1
NM_201536.1 1162 Intron NP_963294.1
NM_201537.1 1162 Intron NP_963831.1
NM_201538.1 1162 Intron NP_963832.1
NM_201539.1 1162 Intron NP_963833.1
NM_201540.1 1162 Intron NP_963834.1
NM_201541.1 1162 Intron NP_963835.1
XM_011536996.2 1162 Intron XP_011535298.1
XM_011536997.1 1162 Intron XP_011535299.1
XM_011536998.1 1162 Intron XP_011535300.1
XM_011536999.1 1162 Intron XP_011535301.1
XM_011537001.1 1162 Intron XP_011535303.1
XM_011537002.1 1162 Intron XP_011535304.1
XM_017021480.1 1162 Intron XP_016876969.1
XM_017021481.1 1162 Intron XP_016876970.1
XM_017021482.1 1162 Intron XP_016876971.1
XM_017021483.1 1162 Intron XP_016876972.1
XM_017021484.1 1162 Intron XP_016876973.1
XM_017021485.1 1162 Intron XP_016876974.1
XM_017021486.1 1162 Intron XP_016876975.1
XM_017021487.1 1162 Intron XP_016876976.1
XM_017021488.1 1162 Intron XP_016876977.1
XM_017021489.1 1162 Intron XP_016876978.1
XM_017021490.1 1162 Intron XP_016876979.1
XM_017021491.1 1162 Intron XP_016876980.1
XM_017021492.1 1162 Intron XP_016876981.1
XM_017021493.1 1162 Intron XP_016876982.1
XM_017021494.1 1162 Intron XP_016876983.1
XM_017021495.1 1162 Intron XP_016876984.1
XM_017021496.1 1162 Intron XP_016876985.1
XM_017021497.1 1162 Intron XP_016876986.1
Gene
RNASE13
Gene Name
ribonuclease A family member 13 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012264.3 1162 UTR 3 NP_001012264.1
Gene
RNASE7
Gene Name
ribonuclease A family member 7
There are no transcripts associated with this gene.

Gene
TPPP2
Gene Name
tubulin polymerization promoting protein family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173846.4 1162 Intron NP_776245.2
XM_005267324.4 1162 Intron XP_005267381.1
XM_011536416.1 1162 Intron XP_011534718.1
XM_011536417.2 1162 Intron XP_011534719.1
XM_011536418.1 1162 Intron XP_011534720.1
XM_011536420.2 1162 Intron XP_011534722.1
XM_017020966.1 1162 Intron XP_016876455.1

View Full Product Details