Product Details

SNP ID
rs78256403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50245347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACTGCTCTCATAAAAATCTGTC[C/T]CTTCTAAGTTATTTCAGTTCTCAGC
Phenotype
MIM: 609584
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
L2HGDH PubMed Links

Gene Details

Gene
L2HGDH
Gene Name
L-2-hydroxyglutarate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024884.2 2262 UTR 3 NP_079160.1
XM_005268075.4 2262 UTR 3 XP_005268132.1
XM_011537166.2 2262 Intron XP_011535468.1
XM_011537167.2 2262 Intron XP_011535469.1
XM_011537168.2 2262 Intron XP_011535470.1
XM_017021655.1 2262 Intron XP_016877144.1
XM_017021656.1 2262 Intron XP_016877145.1
XM_017021657.1 2262 UTR 3 XP_016877146.1
XM_017021658.1 2262 Intron XP_016877147.1

View Full Product Details