Product Details
- SNP ID
-
rs77001619
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:52777226 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCTAATAGTTCAACAGCAGACTC[A/C]AGAAAAGCACAGATGTGATTCTAAC
- Phenotype
-
MIM: 616510
MIM: 615814
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GNPNAT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201557986] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNPNAT1
- Gene Name
- glucosamine-phosphate N-acetyltransferase 1
- Gene
- STYX
- Gene Name
- serine/threonine/tyrosine interacting protein
There are no transcripts associated with this gene.
View Full Product Details