Product Details

SNP ID
rs75766796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:54399973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAACAGACTCCAATTCATATAT[C/T]ATGGTATGTTAGCATTTCTGATCAA
Phenotype
MIM: 123832
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKN3 PubMed Links

Gene Details

Gene
CDKN3
Gene Name
cyclin dependent kinase inhibitor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130851.1 219 Intron NP_001124323.1
NM_005192.3 219 Missense Mutation TCA,TTA S30L NP_005183.2
XM_011536353.2 219 Missense Mutation TCA,TTA S30L XP_011534655.1

View Full Product Details