Product Details

SNP ID
rs78756967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55417897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATCAAACAATATGGGCTTAAGT[A/G]CCAGCTCTACCTCTAACTAGCTTTA
Phenotype
MIM: 613515 MIM: 608964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG14 PubMed Links

Gene Details

Gene
ATG14
Gene Name
autophagy related 14
There are no transcripts associated with this gene.

Gene
TBPL2
Gene Name
TATA-box binding protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199047.2 Intron NP_950248.1

View Full Product Details