Product Details

SNP ID
rs74856420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57205162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTATTAGCACAGTGTGTGAGACTT[A/C]AATCAATACATGTAACTATAATTAT
Phenotype
MIM: 604469
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EXOC5 PubMed Links

Gene Details

Gene
EXOC5
Gene Name
exocyst complex component 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006544.3 5825 UTR 3 NP_006535.1
XM_005267272.3 5825 UTR 3 XP_005267329.1

View Full Product Details