Product Details

SNP ID
rs78534969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59601492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCTAAGAGCTCTTTTAGGGCT[A/G]AGATCATTTCATTTATCTTTATACC
Phenotype
MIM: 600865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RTN1 PubMed Links

Gene Details

Gene
RTN1
Gene Name
reticulon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021136.2 Intron NP_066959.1
NM_206852.2 Intron NP_996734.1
XM_011537063.2 Intron XP_011535365.1
XM_017021578.1 Intron XP_016877067.1

View Full Product Details