Product Details
- SNP ID
-
rs75852319
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:60712276 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCCATTTGTGTGATAAATGTCAT[A/T]TATATTATTTAGTGAAACTTAAGTT
- Phenotype
-
MIM: 606342
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SIX4
PubMed Links
Gene Details
- Gene
- SIX4
- Gene Name
- SIX homeobox 4
View Full Product Details