Product Details

SNP ID
rs78010689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22922544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTCCAAGGTGCAATAGCGGTTG[G/T]TGTCAATCATAGGATCTGTCAGGAA
Phenotype
MIM: 604045
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101926933 PubMed Links

Gene Details

Gene
LOC101926933
Gene Name
uncharacterized LOC101926933
There are no transcripts associated with this gene.

Gene
PRMT5
Gene Name
protein arginine methyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039619.2 1442 Missense Mutation AAC,ACC N515T NP_001034708.1
NM_001282953.1 1442 Missense Mutation AAC,ACC N471T NP_001269882.1
NM_001282954.1 1442 Missense Mutation AAC,ACC N426T NP_001269883.1
NM_001282955.1 1442 Missense Mutation AAC,ACC N488T NP_001269884.1
NM_001282956.1 1442 Missense Mutation AAC,ACC N361T NP_001269885.1
NM_006109.4 1442 Missense Mutation AAC,ACC N532T NP_006100.2
XM_017020927.1 1442 Missense Mutation AAC,ACC N426T XP_016876416.1
Gene
PRMT5-AS1
Gene Name
PRMT5 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RBM23
Gene Name
RNA binding motif protein 23
There are no transcripts associated with this gene.

View Full Product Details