Product Details

SNP ID
rs74373491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70370251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTCACTTTGTGTGTCTCAAG[G/T]CTCTCTCAGGAATCAGTGTCCAAAC
Phenotype
MIM: 609411
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SYNJ2BP PubMed Links

Gene Details

Gene
SYNJ2BP
Gene Name
synaptojanin 2 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018373.2 3305 UTR 3 NP_060843.2
Gene
SYNJ2BP-COX16
Gene Name
SYNJ2BP-COX16 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202547.2 3305 Intron NP_001189476.1
NM_001202548.2 3305 Intron NP_001189477.1
NM_001202549.2 3305 Intron NP_001189478.1

View Full Product Details