Product Details

SNP ID
rs75415848
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24073949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACCCTAGCCCAACTCAAAGTGC[C/T]AACCCTTGCAGACACTCAGAGCATT
Phenotype
MIM: 614716 MIM: 605688 MIM: 162080
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CARMIL3 PubMed Links

Gene Details

Gene
CARMIL3
Gene Name
capping protein regulator and myosin 1 linker 3
There are no transcripts associated with this gene.

Gene
CPNE6
Gene Name
copine 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280558.1 Intron NP_001267487.1
NM_006032.3 Intron NP_006023.1
XM_005268216.1 Intron XP_005268273.1
XM_005268217.1 Intron XP_005268274.1
Gene
NRL
Gene Name
neural retina leucine zipper
There are no transcripts associated with this gene.

View Full Product Details