Product Details

SNP ID
rs74744808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24218169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTAGATGAGCCTCTGCTGTTGT[G/T]GGGGGATTCCCTCTTTCTCCTCCAC
Phenotype
MIM: 610051 MIM: 603171 MIM: 610711
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CHMP4A PubMed Links

Gene Details

Gene
CHMP4A
Gene Name
charged multivesicular body protein 4A
There are no transcripts associated with this gene.

Gene
MDP1
Gene Name
magnesium dependent phosphatase 1
There are no transcripts associated with this gene.

Gene
NEDD8
Gene Name
neural precursor cell expressed, developmentally down-regulated 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006156.2 Intron NP_006147.1
Gene
NEDD8-MDP1
Gene Name
NEDD8-MDP1 readthrough
There are no transcripts associated with this gene.

Gene
TSSK4
Gene Name
testis specific serine kinase 4
There are no transcripts associated with this gene.

View Full Product Details