Product Details

SNP ID
rs77722833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77269250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCCAGGAACTCAGGCGAGGAGA[C/G]ACAGTCCTCTGGGCTGGAGAACTGG
Phenotype
MIM: 605304 MIM: 607439
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR1260A PubMed Links

Gene Details

Gene
MIR1260A
Gene Name
microRNA 1260a
There are no transcripts associated with this gene.

Gene
NGB
Gene Name
neuroglobin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021257.3 541 Missense Mutation CTC,GTC L56V NP_067080.1
Gene
POMT2
Gene Name
protein O-mannosyltransferase 2
There are no transcripts associated with this gene.

Gene
TMEM63C
Gene Name
transmembrane protein 63C
There are no transcripts associated with this gene.

View Full Product Details