Product Details

SNP ID
rs75196191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:80199437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACCCAATGTGAAAAAAGAAAG[A/T]CATGTAAGTTAAGTGACTGTCACTT
Phenotype
MIM: 601413
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DIO2 PubMed Links

Gene Details

Gene
DIO2
Gene Name
deiodinase, iodothyronine, type II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000793.5 4363 UTR 3 NP_000784.2
NM_001007023.4 4363 UTR 3 NP_001007024.1
NM_001242502.2 4363 UTR 3 NP_001229431.1
NM_001242503.2 4363 UTR 3 NP_001229432.1
NM_001324462.1 4363 UTR 3 NP_001311391.1
NM_013989.4 4363 UTR 3 NP_054644.1

View Full Product Details