Product Details

SNP ID
rs79885553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:89803759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGAAGACCCGCCAATTGAAGAT[G/T]TAATGATGACAGCCTCTCCCGCAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB11 PubMed Links

Gene Details

Gene
EFCAB11
Gene Name
EF-hand calcium binding domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284266.1 Intron NP_001271195.1
NM_001284267.1 Intron NP_001271196.1
NM_001284268.1 Intron NP_001271197.1
NM_001284269.1 Intron NP_001271198.1
NM_145231.3 Intron NP_660274.1

View Full Product Details