Product Details

SNP ID
rs76140743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93340971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACATACTGTCTGATTTAACCTCC[A/G]TATCCACATTCTAAGAGAAATATGC
Phenotype
MIM: 610386 MIM: 616855 MIM: 616884
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTBD7 PubMed Links

Gene Details

Gene
BTBD7
Gene Name
BTB domain containing 7
There are no transcripts associated with this gene.

Gene
COX8C
Gene Name
cytochrome c oxidase subunit 8C
There are no transcripts associated with this gene.

Gene
UNC79
Gene Name
unc-79 homolog (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020818.3 Intron NP_065869.3
XM_011537018.2 Intron XP_011535320.1
XM_011537020.2 Intron XP_011535322.1
XM_011537021.2 Intron XP_011535323.1
XM_011537022.2 Intron XP_011535324.1
XM_011537023.2 Intron XP_011535325.1
XM_011537024.2 Intron XP_011535326.1
XM_011537025.2 Intron XP_011535327.1
XM_011537026.2 Intron XP_011535328.1
XM_011537027.2 Intron XP_011535329.1
XM_011537028.2 Intron XP_011535330.1
XM_011537029.2 Intron XP_011535331.1
XM_017021506.1 Intron XP_016876995.1
XM_017021507.1 Intron XP_016876996.1
XM_017021508.1 Intron XP_016876997.1
XM_017021509.1 Intron XP_016876998.1
XM_017021510.1 Intron XP_016876999.1
XM_017021511.1 Intron XP_016877000.1
XM_017021512.1 Intron XP_016877001.1
XM_017021513.1 Intron XP_016877002.1
XM_017021514.1 Intron XP_016877003.1
XM_017021515.1 Intron XP_016877004.1
XM_017021516.1 Intron XP_016877005.1
XM_017021517.1 Intron XP_016877006.1
XM_017021518.1 Intron XP_016877007.1
XM_017021519.1 Intron XP_016877008.1
XM_017021520.1 Intron XP_016877009.1

View Full Product Details