Product Details

SNP ID
rs74536787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94115376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTAGAGCTGTCTCTTTGAGCC[A/G]CTCTGAATGTGCCTCTTACATAATC
Phenotype
MIM: 600009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFI27 PubMed Links

Gene Details

Gene
IFI27
Gene Name
interferon alpha inducible protein 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130080.2 Intron NP_001123552.1
NM_001288952.1 Intron NP_001275881.1
NM_001288954.1 Intron NP_001275883.1
NM_001288956.1 Intron NP_001275885.1
NM_001288957.1 Intron NP_001275886.1
NM_001288958.1 Intron NP_001275887.1
NM_001288959.1 Intron NP_001275888.1
NM_001288960.1 Intron NP_001275889.1
NM_001288995.1 Intron NP_001275924.1
NM_005532.4 Intron NP_005523.3

View Full Product Details