Product Details

SNP ID
rs77719352
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99744290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTCCCAGGGCAGGTCCCTACA[C/T]GGTGACCCAGTGTGATGCTTCCATA
Phenotype
MIM: 602033
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
EML1 PubMed Links

Gene Details

Gene
EML1
Gene Name
echinoderm microtubule associated protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008707.1 Intron NP_001008707.1
NM_004434.2 Intron NP_004425.2
XM_005267397.1 Intron XP_005267454.1
XM_005267398.3 Intron XP_005267455.1
XM_005267399.4 Intron XP_005267456.2
XM_005267400.4 Intron XP_005267457.1
XM_011536540.2 Intron XP_011534842.1
XM_011536541.1 Intron XP_011534843.1
XM_011536542.2 Intron XP_011534844.1
XM_017021074.1 Intron XP_016876563.1

View Full Product Details