Product Details

SNP ID
rs78863423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103697748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCACTGTGCCCATGGGGAGGCCA[C/T]GCCACTCTGCCTGCTGGGTGGATAT
Phenotype
MIM: 600025 MIM: 600675
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLC1 PubMed Links

Gene Details

Gene
KLC1
Gene Name
kinesin light chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130107.1 2471 Intron NP_001123579.1
NM_005552.4 2471 Intron NP_005543.2
NM_182923.3 2471 Intron NP_891553.2
Gene
XRCC3
Gene Name
X-ray repair cross complementing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100118.1 2471 UTR 3 NP_001093588.1
NM_001100119.1 2471 UTR 3 NP_001093589.1
NM_005432.3 2471 UTR 3 NP_005423.1
XM_005268046.2 2471 UTR 3 XP_005268103.1
XM_011537138.2 2471 UTR 3 XP_011535440.1

View Full Product Details