Product Details
- SNP ID
-
rs77381814
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:103699410 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCTGGAAGGCACTGCTCAGCTCA[C/T]GCAGCGTGGCCCCCAGGGACTGCAG
- Phenotype
-
MIM: 600025
MIM: 600675
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KLC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs861539] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLC1
- Gene Name
- kinesin light chain 1
- Gene
- XRCC3
- Gene Name
- X-ray repair cross complementing 3
View Full Product Details