Product Details

SNP ID
rs78564584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:24583374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGAGGACAGTTATATTTTCTTA[A/G]AGAATCACCCCTTTTATCCTTAACT
Phenotype
MIM: 611215
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PWRN1 PubMed Links

Gene Details

Gene
PWRN1
Gene Name
Prader-Willi region non-protein coding RNA 1
There are no transcripts associated with this gene.

View Full Product Details