Product Details
- SNP ID
-
rs75000324
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:150222658 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAGAACCAGCAGCTCCACTCCAC[A/G]GACAGAGTGGATCTCTGCGGCACAG
- Phenotype
-
MIM: 114078
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CAMK2A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140679603] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAMK2A
- Gene Name
- calcium/calmodulin dependent protein kinase II alpha
View Full Product Details