Product Details

SNP ID
rs78564056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141156456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCCATCCAGGGACTTCATTTC[C/T]TTTGAAAATAGCTCAAGATTTGGAC
Phenotype
MIM: 606332
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDHB17P PubMed Links

Gene Details

Gene
PCDHB17P
Gene Name
protocadherin beta 17 pseudogene
There are no transcripts associated with this gene.

Gene
PCDHB6
Gene Name
protocadherin beta 6
There are no transcripts associated with this gene.

View Full Product Details