Product Details

SNP ID
rs75647802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140128569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCTCCCATCTCAGTGCTGGGAA[A/C]TCACCATGTGGAAACCAAGCAAACG
Phenotype
MIM: 600473
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGIP PubMed Links

Gene Details

Gene
IGIP
Gene Name
IgA inducing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007189.1 2634 Missense Mutation AAA,AAC K31N NP_001007190.1
Gene
PURA
Gene Name
purine rich element binding protein A
There are no transcripts associated with this gene.

View Full Product Details