Product Details
- SNP ID
-
rs79408814
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:146447386 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCGGGGACGGGGGCGAGAGTGAA[A/C]GATTCAACCCGGGGGAGCTCAGGTA
- Phenotype
-
MIM: 605409
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TCERG1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6884478] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TCERG1
- Gene Name
- transcription elongation regulator 1
View Full Product Details