Product Details

SNP ID
rs74471144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149150148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCAGCAGCTTCCCAGGGCACAG[C/T]CCAGAGTAGGGAAGCAGGCAGAACG
Phenotype
MIM: 611305
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABLIM3 PubMed Links

Gene Details

Gene
ABLIM3
Gene Name
actin binding LIM protein family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301015.1 Intron NP_001287944.1
NM_001301018.1 Intron NP_001287947.1
NM_001301027.1 Intron NP_001287956.1
NM_001301028.1 Intron NP_001287957.1
NM_014945.3 Intron NP_055760.1
XM_005268392.1 Intron XP_005268449.1
XM_005268393.1 Intron XP_005268450.1
XM_005268394.1 Intron XP_005268451.1
XM_005268395.1 Intron XP_005268452.1
XM_005268396.1 Intron XP_005268453.1
XM_011537609.2 Intron XP_011535911.1
XM_017009238.1 Intron XP_016864727.1

View Full Product Details