Product Details

SNP ID
rs76928805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47736623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGTCTGTTGGGCCTCAACAAG[C/T]GACGGGCTCTCCGCGGAAGCTTCAG
Phenotype
MIM: 606057
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105369748 PubMed Links

Gene Details

Gene
LOC105369748
Gene Name
uncharacterized LOC105369748
There are no transcripts associated with this gene.

Gene
RAPGEF3
Gene Name
Rap guanine nucleotide exchange factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098531.2 4030 UTR 3 NP_001092001.1
NM_001098532.2 4030 UTR 3 NP_001092002.1
NM_006105.5 4030 UTR 3 NP_006096.2
XM_005268571.3 4030 Intron XP_005268628.1
XM_011537752.1 4030 Intron XP_011536054.1
XM_011537755.1 4030 Intron XP_011536057.1
XM_011537758.2 4030 Intron XP_011536060.1
XM_017018688.1 4030 Intron XP_016874177.1

View Full Product Details