Product Details
- SNP ID
-
rs78849694
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48329779 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGAGGCGCCGCCAGCCCCTTCGCA[A/G]GGCGGCCAGGAAGGCCAGAGAAGTG
- Phenotype
-
MIM: 613906
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
H1FNT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1471997] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- H1FNT
- Gene Name
- H1 histone family member N, testis specific
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181788.1 |
800 |
Missense Mutation |
AAG,AGG |
K163R |
NP_861453.1 |
- Gene
- ZNF641
- Gene Name
- zinc finger protein 641
There are no transcripts associated with this gene.
View Full Product Details