Product Details

SNP ID
rs78516105
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52272508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTAACAACTTTATTTTTCAAAA[C/T]AGGTGATAAAGTGAAAAGATAAGCT
Phenotype
MIM: 601928
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KRT86 PubMed Links

Gene Details

Gene
KRT86
Gene Name
keratin 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320198.1 2002 Intron NP_001307127.1
XM_005268866.4 2002 UTR 5 XP_005268923.1
XM_017019296.1 2002 Intron XP_016874785.1

View Full Product Details